missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Methoxyphenyl)propionic acid, 98%

Product Code. 11450244
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11450244 25 g 25g
11460244 100 g 100g
11470244 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11450244 Supplier Thermo Scientific Alfa Aesar Supplier No. A13655.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1929-29-9 | C10H12O3 | 180.203 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1929-29-9
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.203
MDL Number MFCD00002777
InChI Key FIUFLISGGHNPSM-UHFFFAOYSA-N
Synonym 3-4-methoxyphenyl propanoic acid, 3-4-methoxyphenyl propionic acid, benzenepropanoic acid, 4-methoxy, p-methoxyhydrocinnamic acid, 3-p-methoxyphenyl propionic acid, 3-4-methoxy-phenyl-propionic acid, 4-methoxyhydrocinnamic acid, 3-4-methoxyphenyl-propionic acid, 4-methoxyhydro cinnamic acid, hydrocinnamic acid, p-methoxy
PubChem CID 95750
IUPAC Name 3-(4-methoxyphenyl)propanoic acid
SMILES COC1=CC=C(C=C1)CCC(=O)O

Specifications

Melting Point 102°C to 106°C
Quantity 25 g
Beilstein 1953326
Formula Weight 180.2
Percent Purity 98%
Chemical Name or Material 3-(4-Methoxyphenyl)propionic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.