missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Methoxyphenyl)propionyl chloride, 99%

Product Code. 11355429
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11355429 1 g 1g
11365429 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11355429 Supplier Thermo Scientific Alfa Aesar Supplier No. H50416.03
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 15893-42-2 | C10H11ClO2 | 198.65 g/mol

3-(4-Methoxyphenyl)propionyl chloride is an important raw material and intermediate used in pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-(4-Methoxyphenyl)propionyl chloride is an important raw material and intermediate used in pharmaceuticals, agrochemicals and dyestuff.

Solubility
Reacts with water.

Notes
Air & Moisture Sensitive. Protect from humidity and water. Store under dry inert gas away from oxidizing agents and moisture. Incompatible with bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15893-42-2
Molecular Formula C10H11ClO2
Molecular Weight (g/mol) 198.65
MDL Number MFCD02258671
InChI Key FQVJPHCAWYRYCK-UHFFFAOYSA-N
Synonym 3-4-methoxyphenyl propionyl chloride, 3-4-methoxyphenyl propanoyl chloride, acmc-20anjm, p-methoxyhydrocinnamoyl chloride, 4-methoxyphenylpropionyl chloride, 4-methoxydihydrocinnamoyl chloride, 3-4-methoxyphenyl propanyl chloride, benzenepropanoyl chloride, 4-methoxy, 3-4-methoxyphenyl-propionyl chloride, 3-4-methoxy-phenyl-propionyl chloride
PubChem CID 10932441
IUPAC Name 3-(4-methoxyphenyl)propanoyl chloride
SMILES COC1=CC=C(CCC(Cl)=O)C=C1

Specifications

Quantity 1 g
UN Number UN3265
Sensitivity Air and moisture sensitive
Solubility Information Reacts with water.
Formula Weight 198.65
Percent Purity 99%
Chemical Name or Material 3-(4-Methoxyphenyl)propionyl chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.