missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Methyl-1-piperazinylmethyl)benzeneboronic acid pinacol ester, 97%

Product Code. 11412140
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11412140 1 g 1g
11422140 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11412140 Supplier Thermo Scientific Alfa Aesar Supplier No. H28762.03
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 883738-27-0 | C18H29BN2O2 | 316.252 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 883738-27-0
Molecular Formula C18H29BN2O2
Molecular Weight (g/mol) 316.252
MDL Number MFCD09266181
InChI Key PESZAXRJFGLGTR-UHFFFAOYSA-N
Synonym 1-methyl-4-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzyl piperazine, 3-4-methyl-1-piperazinylmethyl benzeneboronic acid pinacol ester, 3-4-methylpiperazin-1-yl methyl phenylboronic acid pinacol ester, 1-methyl-4-3-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methyl piperazine, 1-methyl-4-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methyl piperazine, amtb037, 3-4-methylpiperazin-1-yl methyl phenylboronic acid, pinacol ester, 1-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzyl-4-methylp ip erazine, 1-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzyl-4-methylpiperazine
PubChem CID 46739725
IUPAC Name 1-methyl-4-[[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl]piperazine
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC(=CC=C2)CN3CCN(CC3)C

Specifications

Quantity 1 g
Formula Weight 316.25
Percent Purity 97%
Chemical Name or Material 3-(4-Methyl-1-piperazinylmethyl)benzeneboronic acid pinacol ester

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.