missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Methylbenzoyl)acrylic acid, 98%

Product Code. 11348505
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11348505 5 g 5g
11338505 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11348505 Supplier Thermo Scientific Alfa Aesar Supplier No. L10551.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 20972-36-5 | C11H10O3 | 190.198 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20972-36-5
Molecular Formula C11H10O3
Molecular Weight (g/mol) 190.198
MDL Number MFCD00075338
InChI Key VNJMEFZKYZHWEO-VOTSOKGWSA-N
Synonym 3-4-methylbenzoyl acrylic acid, 2e-4-4-methylphenyl-4-oxobut-2-enoic acid, e-4-oxo-4-p-tolyl but-2-enoic acid, trans-3-4-methylbenzoyl acrylic acid, e-3-p-toluoylacrylic acid, chembl44664, regid_for_cid_691106, 3-4-methyl benzoyl acrylic acid, 3-4-methyl-benzoyl acrylic acid, 4-4-methylphenyl-4-oxo-2-butenoic acid
PubChem CID 691106
IUPAC Name (E)-4-(4-methylphenyl)-4-oxobut-2-enoic acid
SMILES CC1=CC=C(C=C1)C(=O)C=CC(=O)O

Specifications

Melting Point 138°C to 140°C
Quantity 5 g
Beilstein 2691231
Formula Weight 190.2
Percent Purity 98%
Chemical Name or Material 3-(4-Methylbenzoyl)acrylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.