missing translation for 'onlineSavingsMsg'
Learn More

3-[(4-Methylperhydro-1,4-diazepin-1-yl)methyl]benzonitrile, 97%, Thermo Scientific™

Product Code. 11396795
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11396795 5 g Amber Glass Bottle 5g
10402854 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11396795 Supplier Thermo Scientific Maybridge Supplier No. CC55916DE
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 910036-91-8
Molecular Formula C14H19N3
Molecular Weight (g/mol) 229.327
MDL Number MFCD09702379
InChI Key WCQJGGDNUFLTPA-UHFFFAOYSA-N
Synonym 3-4-methylhomopiperazin-1-yl methyl benzonitrile, 3-4-methyl-1,4-diazepan-1-yl methyl benzonitrile, 3-4-methylperhydro-1,4-diazepin-1-yl methyl benzonitrile, 3-4-methyl-1,4-diazaperhydroepinyl methyl benzenecarbonitrile, benzonitrile,3-hexahydro-4-methyl-1h-1,4-diazepin-1-yl methyl
PubChem CID 24229556
IUPAC Name 3-[(4-methyl-1,4-diazepan-1-yl)methyl]benzonitrile
SMILES CN1CCCN(CC1)CC2=CC=CC(=C2)C#N

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 229.33
Percent Purity 97%
Chemical Name or Material 3-[(4-Methylperhydro-1, 4-diazepin-1-yl)methyl]benzonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.