missing translation for 'onlineSavingsMsg'
Learn More

3',5,7-Trihydroxy-4'-methoxyflavanone, 97%

Product Code. 11428897
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11428897 25 g 25g
11408897 1 g 1g
11418897 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11428897 Supplier Thermo Scientific Alfa Aesar Supplier No. B20528.14
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 520-33-2 | C16H14O6 | 302.28 g/mol

(±)-Hesperetin is an antioxidant flavonoid. Induces G1-phase cell cycle arrest. Anti-inflammatory. (±)-Hesperetin suppresses NF-κB activation. Reduces cholesterol biosynthesis. Inhibits lipid peroxidation. Neuroprotective against neuronal oxidative damage.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(±)-Hesperetin is an antioxidant flavonoid. Induces G1-phase cell cycle arrest. Anti-inflammatory. (±)-Hesperetin suppresses NF-κB activation. Reduces cholesterol biosynthesis. Inhibits lipid peroxidation. Neuroprotective against neuronal oxidative damage.

Solubility
Soluble in water (partly), dilute alkalis, and ethanol (50 mg/ml).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 520-33-2
Molecular Formula C16H14O6
Molecular Weight (g/mol) 302.28
MDL Number MFCD00075646
InChI Key AIONOLUJZLIMTK-AWEZNQCLSA-N
Synonym hesperetin, hesperitin, 3',5,7-trihydroxy-4'-methoxyflavanone, hesperin, yso2, cyanidanon 4'-methyl ether 1626, 5,7,3'-trihydroxy-4'-methoxyflavanone, --hesperetin, prestwick_908, unii-q9q3d557f1
PubChem CID 72281
ChEBI CHEBI:28230
IUPAC Name (2S)-5,7-dihydroxy-2-(3-hydroxy-4-methoxyphenyl)-2,3-dihydrochromen-4-one
SMILES COC1=CC=C(C=C1O)[C@@H]1CC(=O)C2=C(O)C=C(O)C=C2O1

Specifications

Melting Point 227°C to 232°C
Quantity 25 g
Beilstein 309850
Merck Index 14,4670
Solubility Information Soluble in water (partly),dilute alkalis,and ethanol (50mg/ml).
Formula Weight 302.3
Percent Purity 97%
Chemical Name or Material 3',5,7-Trihydroxy-4'-methoxyflavanone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.