missing translation for 'onlineSavingsMsg'
Learn More

3,5,7-Trihydroxyflavone, 97%

Product Code. 11329055
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11329055 25 mg 25mg
11339055 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11329055 Supplier Thermo Scientific Alfa Aesar Supplier No. L14223.MB
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 548-83-4 | C15H10O5 | 270.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 548-83-4
Molecular Formula C15H10O5
Molecular Weight (g/mol) 270.24
MDL Number MFCD00006833
InChI Key VCCRNZQBSJXYJD-UHFFFAOYSA-N
Synonym galangin, norizalpinin, 3,5,7-trihydroxyflavone, 3,5,7-trihydroxy-2-phenyl-4h-chromen-4-one, 3,5,7-trioh-flavone, unii-142fwe6ecs, 3,5,7-trihydroxy-2-phenyl-4-benzopyrone, 4h-1-benzopyran-4-one, 3,5,7-trihydroxy-2-phenyl, flavone, 3,5,7-trihydroxy, 4h-benzopyran-4-one, 3,5,7-trihydroxy-2-phenyl
PubChem CID 5281616
ChEBI CHEBI:5262
IUPAC Name 3,5,7-trihydroxy-2-phenylchromen-4-one
SMILES C1=CC=C(C=C1)C2=C(C(=O)C3=C(C=C(C=C3O2)O)O)O

Specifications

Melting Point 214°C to 216°C
Quantity 25 mg
Beilstein 272179
Merck Index 14,4339
Formula Weight 270.24
Percent Purity 97%
Chemical Name or Material 3,5,7-Trihydroxyflavone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.