missing translation for 'onlineSavingsMsg'
Learn More

3,5-Bis(trifluoromethyl)aniline, 98%

Product Code. 11408156
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11408156 50 g 50g
11418156 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11408156 Supplier Thermo Scientific Alfa Aesar Supplier No. A15289.18
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 328-74-5 | C8H5F6N | 229.125 g/mol

3,5-Bis(trifluoromethyl)aniline was used in the synthesis of N-[2-hydroxy-1-naphthylidene]-3, 5-bis(trifluoromethyl)aniline, Schiff?s base and 5,7-bis(trifluoromethyl)aniline. It was also used in the synthesis of N-1-phenylethyl-3,5-bis(trifluoromethyl)aniline via titanium-catalyzed hydroamination reaction and N,N?-bis[3,5-bis(tri-fluoromethyl)phenyl]thiourea, organocatalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Bis(trifluoromethyl)aniline was used in the synthesis of N-[2-hydroxy-1-naphthylidene]-3, 5-bis(trifluoromethyl)aniline, Schiff′s base and 5,7-bis(trifluoromethyl)aniline. It was also used in the synthesis of N-1-phenylethyl-3,5-bis(trifluoromethyl)aniline via titanium-catalyzed hydroamination reaction and N,N′-bis[3,5-bis(tri-fluoromethyl)phenyl]thiourea, organocatalyst.

Solubility
Not miscible in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 328-74-5
Molecular Formula C8H5F6N
Molecular Weight (g/mol) 229.125
MDL Number MFCD00000394
InChI Key CDIDGWDGQGVCIB-UHFFFAOYSA-N
Synonym 3,5-bis trifluoromethyl aniline, 3,5-di trifluoromethyl aniline, benzenamine, 3,5-bis trifluoromethyl, 3,5-bis trifluoromethyl benzenamine, 3',5'-bis-trifluoromethyl aniline, 3,5-bis-trifluoromethylaniline, alpha,alpha,alpha,alpha',alpha',alpha'-hexafluoro-3,5-xylidine, 3,5-bis-trifluoromethyl-phenylamine, aniline, 3,5-bis trifluoromethyl
PubChem CID 9480
IUPAC Name 3,5-bis(trifluoromethyl)aniline
SMILES C1=C(C=C(C=C1C(F)(F)F)N)C(F)(F)F

Specifications

Density 1.473
Boiling Point 75°C to 78°C (10 mmHg)
Flash Point 83°C (181°F)
Odor Recognizable
Refractive Index 1.434
Quantity 50 g
UN Number UN2810
Beilstein 654318
Solubility Information Not miscible in water.
Formula Weight 229.13
Percent Purity 98%
Chemical Name or Material 3,5-Bis(trifluoromethyl)aniline
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.