missing translation for 'onlineSavingsMsg'
Learn More

3,5-Bis(trifluoromethyl)benzeneboronic acid, 97+%

Product Code. 11450183
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11450183

Brand: Thermo Scientific Alfa Aesar A11373.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 73852-19-4 | C8H5BF6O2 | 257.93 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 73852-19-4
Molecular Formula C8H5BF6O2
Molecular Weight (g/mol) 257.93
MDL Number MFCD00051850
InChI Key BPTABBGLHGBJQR-UHFFFAOYSA-N
Synonym 3,5-bis trifluoromethyl phenylboronic acid, 3,5-bis trifluoromethyl benzeneboronic acid, 3,5-bis trifluoromethyl phenyl boronic acid, 3,5-bis-trifluoromethylphenylboronic acid, 3,5-bis trifluoromethylphenyl boronic acid, 3,5-bis trifluoromethyl phenylboroic acid, btfpba, 3,5-bis trifluoromethyl phenyl boranediol
PubChem CID 156265
IUPAC Name [3,5-bis(trifluoromethyl)phenyl]boronic acid
SMILES OB(O)C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F

Specifications

Melting Point 219°C to 224°C
Quantity 1 g
Beilstein 7379990
Formula Weight 257.93
Percent Purity ≥97%
Chemical Name or Material 3,5-Bis (trifluoromethyl)benzeneboronic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.