missing translation for 'onlineSavingsMsg'
Learn More

3,5-Bis(trifluoromethyl)benzenecarboximidamide hydrochloride, ≥95%, Thermo Scientific™

Product Code. 10136802
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10136802 1 g Amber Glass Bottle 1g
10320783 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10136802 Supplier Thermo Scientific Maybridge Supplier No. RF00749DA
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 97603-94-6
Molecular Formula C9H7ClF6N2
Molecular Weight (g/mol) 292.609
MDL Number MFCD00276604
InChI Key GOCQSQHVIVLYOG-UHFFFAOYSA-N
Synonym 3,5-bis trifluoromethyl benzamidine hydrochloride, 3,5-bis trifluoromethyl benzenecarboximidamide hydrochloride, acmc-20andy, 3,5-bis trifluoromethyl benzimidamide hydrochloride, 3,5-bis trifluoromethyl benzenecarboxamidine, chloride, 3,5-bis trifluoromethyl benzene-1-carboximidamide-hydrogen chloride 1/1
PubChem CID 13590400
IUPAC Name 3,5-bis(trifluoromethyl)benzenecarboximidamide;hydrochloride
SMILES C1=C(C=C(C=C1C(F)(F)F)C(F)(F)F)C(=N)N.Cl

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 292.61
Percent Purity 95%
Chemical Name or Material 3, 5-Bis(trifluoromethyl)benzamidine hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.