missing translation for 'onlineSavingsMsg'
Learn More

3,5-Bis(trifluoromethyl)benzylamine, 95%

Product Code. 10531151 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10531151 1 g Glass bottle 1g
10167993 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10531151 Supplier Thermo Scientific Acros Supplier No. 305640010
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 85068-29-7 | C9H7F6N | 243.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 85068-29-7
Molecular Formula C9H7F6N
Molecular Weight (g/mol) 243.15
MDL Number MFCD00009909
InChI Key DHVHORCFFOSRBP-UHFFFAOYSA-N
Synonym 3,5-bis trifluoromethyl benzylamine, 3,5-bis trifluoromethyl phenyl methanamine, 3,5-di trifluoromethyl benzylamine, 3,5-ditrifluoromethylbenzylamine, mbt-bym, 3,5-bis trifluoromethyl phenyl methylamine, benzenemethanamine, 3,5-bis trifluoromethyl, 3,5-bis-trifluoromethyl-benzylamine
PubChem CID 521099
IUPAC Name [3,5-bis(trifluoromethyl)phenyl]methanamine
SMILES C1=C(C=C(C=C1C(F)(F)F)C(F)(F)F)CN

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 46°C to 55°C
Color White to Beige
Boiling Point 82°C to 84°C (15.0 mmHg)
Flash Point 78°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula (CF3)2C6H3CH2NH2
Quantity 1 g
Formula Weight 243.15
Percent Purity 95%
Physical Form Crystalline Mass, Chunks or Crystals and Powder
Chemical Name or Material 3, 5-Bis(trifluoromethyl)benzylamine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.