missing translation for 'onlineSavingsMsg'
Learn More

3,5-Bis(trifluoromethyl)phenol, 97%

Product Code. 11459482
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11459482 1 g 1g
11469482 5 g 5g
11479482 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11459482 Supplier Thermo Scientific Alfa Aesar Supplier No. A10183.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 349-58-6 | C8H4F6O | 230.109 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 349-58-6
Molecular Formula C8H4F6O
Molecular Weight (g/mol) 230.109
MDL Number MFCD00000386
InChI Key ODSXJQYJADZFJX-UHFFFAOYSA-N
Synonym 3,5-bis trifluoromethyl phenol, phenol, 3,5-bis trifluoromethyl, 3,5-di trifluoromethyl phenol, 3,5-bis-trifluoromethylphenol, 3,5-ditrifluoromethylphenol, 3,5-bis-trifluoromethyl phenol, alpha,alpha,alpha,alpha',alpha',alpha'-hexafluoro-3,5-xylenol, pubchem12483, mbt-oh, acmc-1clsr
PubChem CID 67680
IUPAC Name 3,5-bis(trifluoromethyl)phenol
SMILES C1=C(C=C(C=C1C(F)(F)F)O)C(F)(F)F

Specifications

Melting Point 20°C to 21°C
Density 1.511
Boiling Point 97°C to 98°C (50 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.415
Quantity 1 g
Beilstein 2123670
Formula Weight 230.11
Percent Purity 97%
Chemical Name or Material 3,5-Bis (trifluoromethyl)phenol
compliance-icons
Product Identifier
  • 3,5-Bis(trifluoromethyl)phenol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: 3,5-Bis(trifluoromethyl)phenol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.