missing translation for 'onlineSavingsMsg'
Learn More

3,5-Bis(trifluoromethyl)styrene, 96%, stab.

Product Code. 11445507
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11445507 1 g 1g
11455507 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11445507 Supplier Thermo Scientific Alfa Aesar Supplier No. B23499.03
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 349-59-7 | C10H6F6 | 240.15 g/mol

3,5-Bis(trifluoromethyl)styrene is used to produce other chemicals for example 1-(3, 5-bis-Trifluoromethyl-phenyl)-ethane-1, 2-diol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Bis(trifluoromethyl)styrene is used to produce other chemicals for example 1-(3, 5-bis-Trifluoromethyl-phenyl)-ethane-1, 2-diol.

Solubility
Sparingly soluble in water.

Notes
Store in a cool, dry conditions, in well sealed conditions. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 349-59-7
Molecular Formula C10H6F6
Molecular Weight (g/mol) 240.15
MDL Number MFCD00075538
InChI Key LFICVUCVPKKPFF-UHFFFAOYSA-N
Synonym 3,5-bis trifluoromethyl styrene, 1,3-bis trifluoromethyl-5-vinylbenzene, 1-ethenyl-3,5-bis trifluoromethyl benzene, 3,5-bis trifluoromethyl styrene, stab. with 4-methoxyphenol, pubchem2736, acmc-1cmtq, 3,5-bis trifluromethyl styrene, 1,3-bis-trifluoromethyl-5-vinyl-benzene, 3,5-bis trifluoromethyl styrene, stab.
PubChem CID 2773242
IUPAC Name 1-ethenyl-3,5-bis(trifluoromethyl)benzene
SMILES FC(F)(F)C1=CC(=CC(C=C)=C1)C(F)(F)F

Specifications

Melting Point 7°C to 8°C
Density 1.334
Boiling Point 59°C to 61°C (20 mmHg)
Flash Point 50°C (122°F)
Refractive Index 1.425
Quantity 1 g
UN Number UN1993
Solubility Information Sparingly soluble in water.
Formula Weight 240.15
Percent Purity 98%
Chemical Name or Material 3,5-Bis(trifluoromethyl)styrene, stabilized
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.