missing translation for 'onlineSavingsMsg'
Learn More

3,5-Bis(trifluoromethyl)thiophenol, 98%

Product Code. 11407646
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11407646

Brand: Thermo Scientific Alfa Aesar A14669.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 130783-02-7 | C8H4F6S | 246.17 g/mol

3,5-Bis(trifluoromethyl)thiophenol, is an important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Bis(trifluoromethyl)thiophenol, is an important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Insoluble in water but soluble in alcohol and ether.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents. Air Sensitive.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 130783-02-7
Molecular Formula C8H4F6S
Molecular Weight (g/mol) 246.17
MDL Number MFCD00042273
InChI Key KCAQWPZIMLLEAF-UHFFFAOYSA-N
Synonym 3,5-bis trifluoromethyl benzenethiol, 3,5-bis trifluoromethyl thiophenol, 3,5-bis trifluoromethyl benzene-1-thiol, pubchem15275, acmc-209bji, 3,5-bis trifluoromethyl-benzenethiol, 3,5-bis trifluoromethyl benzenethiol #
PubChem CID 518690
IUPAC Name 3,5-bis(trifluoromethyl)benzenethiol
SMILES C1=C(C=C(C=C1C(F)(F)F)S)C(F)(F)F

Specifications

Boiling Point 63°C to 64°C (15 mmHg)
Quantity 5 g
UN Number UN2810
Sensitivity Air Sensitive
Solubility Information Insoluble in water but soluble in alcohol and ether.
Formula Weight 246.18
Percent Purity 98%
Chemical Name or Material 3,5-Bis(trifluoromethyl)thiophenol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.