missing translation for 'onlineSavingsMsg'
Learn More

3,5-Di-O-(p-chlorobenzoyl)-2-deoxy-α-D-ribofuranosyl Chloride, TRC

Product Code. 30006615
Change view
Click to view available options
Quantity:
1 g
2.5 g
Unit Size:
1g
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30006615 1 g 1g
30012634 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30006615 Supplier Toronto Research Chemicals Supplier No. TRCD4322351G
 Restricted Item
Estimated Shipment: 30-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21740-23-8
Molecular Formula C19 H15 Cl3 O5
Molecular Weight (g/mol) 429.6784
Synonym 2-Deoxy-α-D-erythro-pentofuranosyl Chloride 3,5-Bis(4-chlorobenzoate), 1-Chloro-2-deoxy-3,5-O-bis(p-chlorobenzoyl)-α-D-erythro-pentofuranose, 3,5-Bis(4-chlorobenzoyl)-2-deoxy-.alpha.-D-ribofuranosyl Chloride, NSC 140594, 3,5-Di-O-(p-chlorobenzoyl)-2-deoxy-α-D-ribofuranosyl Chloride
IUPAC Name [(2R,3S)-5-chloro-3-(4-chlorobenzoyl)oxyoxolan-2-yl]methyl 4-chlorobenzoate
SMILES ClC1C[C@H](OC(=O)c2ccc(Cl)cc2)[C@@H](COC(=O)c3ccc(Cl)cc3)O1

Specifications

Quantity 1 g
InChI Formula InChI=1S/C19H15Cl3O5/c20-13-5-1-11(2-6-13)18(23)25-10-16-15(9-17(22)26-16)27-19(24)12-3-7-14(21)8-4-12/h1-8,15-17H,9-10H2/t15-,16+,17?/m0/s1
Formula Weight 427.9985
Chemical Name or Material 3,5-Di-O-(p-chlorobenzoyl)-2-deoxy-alpha-D-ribofuranosyl Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.