missing translation for 'onlineSavingsMsg'
Learn More

3,5-Di(tert-butyl)-4-hydroxybenzonitrile, 97%, Thermo Scientific™

Product Code. 10140672
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10140672 1 g 1g
10110772 10 g 10g
10031614 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10140672 Supplier Thermo Scientific Maybridge Supplier No. BTB09888DA
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 1988-88-1
Molecular Formula C15H21NO
Molecular Weight (g/mol) 231.34
MDL Number MFCD00156137
InChI Key AKXIIOLURNATOC-UHFFFAOYSA-N
Synonym 3,5-di-tert-butyl-4-hydroxybenzonitrile, 3,5-di tert-butyl-4-hydroxybenzonitrile, benzonitrile, 3,5-bis 1,1-dimethylethyl-4-hydroxy, benzonitrile, 3,5-di-tert-butyl-4-hydroxy, 4-cyano-2,6-di-tert-butylphenol, 3,5-bis tert-butyl-4-hydroxybenzenecarbonitrile, acmc-20a6ry, 2,6-bis tert-butyl-4-cyanophenol, 3,5-di-t-butyl-4-hydroxybenzonitrile, 4-hydroxy-3,5-di-t-butylbenzonitrile
PubChem CID 137274
SMILES CC(C)(C)C1=CC(=CC(=C1O)C(C)(C)C)C#N

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
IUPAC Name 3,5-ditert-butyl-4-hydroxybenzonitrile
Formula Weight 231.34
Percent Purity 97%
Chemical Name or Material 3, 5-Di(tert-butyl)-4-hydroxybenzonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.