missing translation for 'onlineSavingsMsg'
Learn More

3,5-di-Tert-butyl-4-hydroxybenzyl Alcohol, TRC

Product Code. 30179041
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
Unit Size:
1g
500mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30179041 1 g 1g
30178934 500 mg 500mg
30179585 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30179041 Supplier Toronto Research Chemicals Supplier No. TRCD1781551G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 88-26-6
Molecular Formula C15 H24 O2
Molecular Weight (g/mol) 236.35
Synonym Benzyl alcohol, 3,5-di-tert-butyl-4-hydroxy- (6CI,7CI,8CI), 3,5-Bis(1,1-dimethylethyl)-4-hydroxybenzenemethanol, 2,6-Di-tert-butyl-4-hydroxymethylphenol, 3,5-Di-t-butyl-4-hidroxy-benzyl alcohol, 3,5-Di-tert-butyl-4-hydroxybenzyl alcohol, 3,5-Di-tert-butyl-4-hydroxyphenylmethanol, 4-Hydroxy-3,5-di-tert-butylbenzyl alcohol, 4-Hydroxymethyl-2,6-di-tert-butylphenol, AO 754, Ethanox 754, Ionox 100, NSC 159133
IUPAC Name 2,6-ditert-butyl-4-(hydroxymethyl)phenol
SMILES CC(C)(C)C1=CC(=CC(=C1O)C(C)(C)C)CO

Specifications

Quantity 1 g
InChI Formula InChI=1S/C15H24O2/c1-14(2,3)11-7-10(9-16)8-12(13(11)17)15(4,5)6/h7-8,16-17H,9H2,1-6H3
Formula Weight 236.1776
Chemical Name or Material 2,6-Di-tert-butyl-4-hydroxymethylphenol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.