missing translation for 'onlineSavingsMsg'
Learn More

3,5-Di-tert-butyl-4-hydroxyphenylpropionic acid, 98%

Product Code. 11454347
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11454347 100 g 100g
11434347 5 g 5g
11444347 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11454347 Supplier Thermo Scientific Alfa Aesar Supplier No. B20412.22
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 20170-32-5 | C17H26O3 | 278.392 g/mol

It is employed as an active pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as an active pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20170-32-5
MDL Number MFCD00017519
Synonym 3-3,5-di-tert-butyl-4-hydroxyphenyl propionic acid, 3-3,5-di-tert-butyl-4-hydroxyphenyl propanoic acid, fenozan, 3,5-di-tert-butyl-4-hydroxyphenylpropionic acid, benzenepropanoic acid, 3,5-bis 1,1-dimethylethyl-4-hydroxy, unii-x6g09g700f, 3,5-di-tert-butyl-4-hydroxyhydocinnamic acid, 3,5-bis 1,1-dimethylethyl-4-hydroxybenzenepropanoic acid, 3-3,5-ditertbutyl-4-hydroxyphenyl propionic acid, 3-3,5-bis tert-butyl-4-hydroxyphenyl propanoic acid
PubChem CID 88389
IUPAC Name 3-(3,5-ditert-butyl-4-hydroxyphenyl)propanoic acid

Specifications

Melting Point 172°C to 175°C
Molecular Formula C17H25O3
Quantity 100 g
Solubility Information Slightly soluble aq. soln..
InChI Key WPMYUUITDBHVQZ-UHFFFAOYSA-M
SMILES CC(C)(C)C1=CC(CCC([O-])=O)=CC(=C1O)C(C)(C)C
Molecular Weight (g/mol) 277.39
Formula Weight 278.4
Percent Purity 98%
Chemical Name or Material 3,5-Di-tert-butyl-4-hydroxyphenylpropionic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.