missing translation for 'onlineSavingsMsg'
Learn More

3,5-Di-tert-butyl-4-hydroxyphenylpropionic Acid, TRC

Product Code. 30133525
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30133525 25 g 25g
30007564 1 g 1g
30012262 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133525 Supplier Toronto Research Chemicals Supplier No. TRCD41755525G
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20170-32-5
Molecular Formula C17H26O3
Molecular Weight (g/mol) 278.39
Synonym Benzenepropanoic acid, 3,5-bis(1,1-dimethylethyl)-4-hydroxy- (9CI, ACI), 3,5-Bis(1,1-dimethylethyl)-4-hydroxybenzenepropanoic acid (ACI), Hydrocinnamic acid, 3,5-di-tert-butyl-4-hydroxy- (6CI, 7CI, 8CI), 3,5-Di-tert-butyl-4-hydroxybenzenepropanoic acid, 3,5-Di-tert-butyl-4-hydroxyhydrocinnamic acid, 3,5-Di-tert-butyl-4-hydroxyphenylpropionic acid, 3-(3,5-Di-tert-butyl-4-hydroxyphenyl)propanoic acid, 3-(3,5-Di-tert-butyl-4-hydroxyphenyl)propionic acid, 3-(3',5'-Di-tert-butyl-4'-hydroxyphenyl)propionic acid, 3-(3'5'-Di-tert-butyl-4'-hydroxyphenyl) propionic acid, 4-Hydroxy-3,5-di-tert-butylphenylpropionic acid, Antioxidant AO, Fenoksan, Fenoxan, Fenozan, Fenozan acid, Irganox 1310, Irganox B 2, Phenosan, Phenoxan, Phenoxan (antioxidant), Phenozan, β-(3,5-Di-tert-butyl-4-hydroxyphenyl)propionic acid, β-(4-Hydroxy-3,5-di-tert-butylphenyl)propanoic acid, β-(4-Hydroxy-3,5-di-tert-butylphenyl)propionic acid, 3,5-Di-tert-butyl-4-hydroxyhydrocinnamic Acid, 3,5-Bis(1,1-dimethylethyl)-4-hydroxybenzenepropanoic Acid, 3,5-Di-tert-butyl-4-hydroxybenzenepropanoic Acid, 3,5-Di-tert-butyl-4-hydroxyhydrocinnamic Acid, 3,5-Di-tert-butyl-4-hydroxyphenylpropionic Acid, 3-(3,5-Di-tert-butyl-4-hydroxyphenyl)propanoic Acid, 3-(3,5-Di-tert-butyl-4-hydroxyphenyl)propionic Acid, 3-(3',5'-Di-tert-butyl-4'-hydroxyphenyl)propionic Acid, 4-Hydroxy-3,5-di-tert-butylphenylpropionic Acid, Fenoksan, Fenoxan, Fenozan Fenozan Acid, Irganox 1310, Irganox B 2, Phenosan, Phenoxan, Phenoxan (antioxidant), Phenozan, β-(3,5-Di-tert-butyl-4-hydroxyphenyl)propionic Acid β-(4-Hydroxy-3,5-di-tert-butylphenyl)propanoic Acid, β-(4-Hydroxy-3,5-di-tert-butylphenyl)propionic Acid
IUPAC Name 3-(3,5-ditert-butyl-4-hydroxyphenyl)propanoic acid
SMILES CC(C)(C)c1cc(CCC(=O)O)cc(c1O)C(C)(C)C

Specifications

Quantity 25 g
InChI Formula InChI=1S/C17H26O3/c1-16(2,3)12-9-11(7-8-14(18)19)10-13(15(12)20)17(4,5)6/h9-10,20H,7-8H2,1-6H3,(H,18,19)
Formula Weight 278.1882
Percent Purity >95%
Chemical Name or Material 3,5-Di-tert-butyl-4-hydroxyphenylpropionic Acid
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.