missing translation for 'onlineSavingsMsg'
Learn More

3,5-Di-tert-butylcatechol, 99%, Thermo Scientific Chemicals

Product Code. 10313532 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10313532 5 g Glass bottle 5g
10060910 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10313532 Supplier Thermo Scientific Acros Supplier No. 112970050
 Restricted Item
Estimated Shipment: 09-09-2026
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1020-31-1
Molecular Formula C14H22O2
Molecular Weight (g/mol) 222.33
MDL Number MFCD00008819
InChI Key PJZLSMMERMMQBJ-UHFFFAOYSA-N
Synonym 3,5-di-tert-butylcatechol, 3,5-di-t-butylcatechol, 3,5-di-tert-butylbenzene-1,2-diol, 3,5-di-tert-butylpyrocatechol, 1,2-benzenediol, 3,5-bis 1,1-dimethylethyl, 4,6-di-tert-butylpyrocatechol, unii-1328kn3f8b, 1,2-benzenediol, 3,5-di 1,1-dimethylethyl, pyrocatechol, 3,5-di-tert-butyl, 3,5-bis tert-butyl benzene-1,2-diol
PubChem CID 66099
IUPAC Name 3,5-ditert-butylbenzene-1,2-diol
SMILES CC(C)(C)C1=CC(=C(C(=C1)O)O)C(C)(C)C

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 97°C to 100°C
Color White to Beige
Boiling Point >1000°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula [(CH3)3C]2C6H2(OH)2
Quantity 5 g
Formula Weight 222.33
Percent Purity 99%
Grade Technical
Physical Form Needle-like Crystalline Powder
Chemical Name or Material 3, 5-Di-tert-butylcatechol, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.