missing translation for 'onlineSavingsMsg'
Learn More

3,5-Di-tert-butylsalicylic acid, 97%

Product Code. 15491248
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15491248 5 g 5g
15401258 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15491248 Supplier Thermo Scientific Alfa Aesar Supplier No. H55102.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 19715-19-6 | C15H22O3 | 250.338 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19715-19-6
Molecular Formula C15H22O3
Molecular Weight (g/mol) 250.338
MDL Number MFCD00059058
InChI Key ZWQBZEFLFSFEOS-UHFFFAOYSA-N
Synonym 3,5-di-tert-butyl-2-hydroxybenzoic acid, 3,5-bis-tert-butylsalicylic acid, 3,5-di-tert-butylsalicylic acid, benzoic acid, 3,5-bis 1,1-dimethylethyl hydroxy, benzoic acid, 3,5-bis 1,1-dimethylethyl-2-hydroxy, 3,5-ditertbutyl salicylic acid, dibutylsalicylicacid, acmc-1c7qv, 3,2-bis 1,1-dimethylethyl-2-hydroxybenzoic acid, ksc466o9n
PubChem CID 88208
IUPAC Name 3,5-ditert-butyl-2-hydroxybenzoic acid
SMILES CC(C)(C)C1=CC(=C(C(=C1)C(=O)O)O)C(C)(C)C

Specifications

Melting Point 157°C to 162°C
Flash Point 179°C (354°F)
Quantity 5 g
Formula Weight 250.34
Percent Purity 97%
Chemical Name or Material 3,5-Di-tert-butylsalicylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.