missing translation for 'onlineSavingsMsg'
Learn More

3,5-Di(trifluoromethyl)aniline, 97%, Maybridge

Product Code. 10746735
Change view
Click to view available options
Quantity:
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10746735 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10746735 Supplier Thermo Scientific Maybridge Supplier No. SB00812EA
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 328-74-5
CAS Min % 97.0
Density 1.47g/mL
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C8H5F6N
Linear Formula (CF3)2C6H3NH2
MDL Number MFCD00000394
Quantity 10 g
Synonym 3,5-bis trifluoromethyl aniline, 3,5-di trifluoromethyl aniline, benzenamine, 3,5-bis trifluoromethyl, 3,5-bis trifluoromethyl benzenamine, 3',5'-bis-trifluoromethyl aniline, 3,5-bis-trifluoromethylaniline, alpha,alpha,alpha,alpha',alpha',alpha'-hexafluoro-3,5-xylidine, 3,5-bis-trifluoromethyl-phenylamine, aniline, 3,5-bis trifluoromethyl
InChI Key CDIDGWDGQGVCIB-UHFFFAOYSA-N
SMILES C1=C(C=C(C=C1C(F)(F)F)N)C(F)(F)F
IUPAC Name 3,5-bis(trifluoromethyl)aniline
Molecular Weight (g/mol) 229.125
PubChem CID 9480
Formula Weight 229.12
Percent Purity 97%
Chemical Name or Material 3, 5-Di(trifluoromethyl)aniline
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.