missing translation for 'onlineSavingsMsg'
Learn More

3,5-Di(trifluoromethyl)phenol, 97%, Maybridge

Product Code. 10002174
Change view
Click to view available options
Quantity:
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10002174 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10002174 Supplier Thermo Scientific Maybridge Supplier No. SB01119EA

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 349-58-6
CAS Min % 97.0
Density 1.51g/mL
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C8H4F6O
MDL Number MFCD00000386
Quantity 10 g
Synonym 3,5-bis trifluoromethyl phenol, phenol, 3,5-bis trifluoromethyl, 3,5-di trifluoromethyl phenol, 3,5-bis-trifluoromethylphenol, 3,5-ditrifluoromethylphenol, 3,5-bis-trifluoromethyl phenol, alpha,alpha,alpha,alpha',alpha',alpha'-hexafluoro-3,5-xylenol, pubchem12483, mbt-oh, acmc-1clsr
InChI Key ODSXJQYJADZFJX-UHFFFAOYSA-N
SMILES C1=C(C=C(C=C1C(F)(F)F)O)C(F)(F)F
IUPAC Name 3,5-bis(trifluoromethyl)phenol
Molecular Weight (g/mol) 230.109
PubChem CID 67680
Formula Weight 230.11
Percent Purity 97%
Chemical Name or Material 3, 5-Di(trifluoromethyl)phenol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.