missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dibromo-4-methylaniline, 99%

Product Code. p-7046089
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11357886

Brand: Thermo Scientific Alfa Aesar L13101.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13194-73-5 | C7H7Br2N | 264.948 g/mol

3,5-Dibromo-4-methylaniline is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Dibromo-4-methylaniline is used as pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13194-73-5
Molecular Formula C7H7Br2N
Molecular Weight (g/mol) 264.948
MDL Number MFCD00151806
InChI Key AQZDIKCNODUMNY-UHFFFAOYSA-N
PubChem CID 7015779
IUPAC Name 3,5-dibromo-4-methylaniline
SMILES CC1=C(C=C(C=C1Br)N)Br

Specifications

Melting Point 90°C to 92°C
Quantity 1 g
Beilstein 2639285
Synonym 3,5-dibromo-p-toluidine, 4-amino-2,6-dibromotoluene, benzenamine, 3,5-dibromo-4-methyl, acmc-1bt4r, ksc495c9l, 3,5-dibrom-4-methyl-anilin, 3,5-dibromo-4-methylphenylamine, 3,5-dibromo-4-methylphenyl amine, benzenamine,3,5-dibromo-4-methyl, 3,5-bis bromanyl-4-methyl-aniline
Solubility Information Insoluble in water.
Formula Weight 264.96
Percent Purity 99%
Chemical Name or Material 3,5-Dibromo-4-methylaniline

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.