missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dibromo-o-phenylenediamine monohydrochloride, 99%, Thermo Scientific™

Product Code. 10677382 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mg
500 mg
Packaging:
Glass bottle
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10677382 500 mg Glass bottle 500mg
10348360 100 mg Glass bottle 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10677382 Supplier Thermo Scientific Acros Supplier No. 223735000
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 75568-11-5
Molecular Formula C6H6Br2N2·HCl
Molecular Weight (g/mol) 302.4
MDL Number MFCD00012967
InChI Key QOZDVKFQMGARCC-UHFFFAOYSA-N
Synonym 3,5-dibromo-1,2-phenylenediamine monohydrochloride, 3,5-dibromobenzene-1,2-diamine hydrochloride, acmc-20andn, 1,2-diamino-3,5-dibromobenzene monohydrochloride, 3,5-bis bromanyl benzene-1,2-diamine hydrochloride, 3,5-dibromo-1,2-phenylenediamine hydrochloride, 1,2-benzenediamine,3,5-dibromo-, hydrochloride 1:1, 3,5-dibromo-ortho-phenylenediamine monohydrochloride, 3,5-dibromobenzene-1,2-diamine-hydrogen chloride 1/1, 3,5-dibromo-1,2-phenylenediaminemonohydrochloride sensitivereagentforthedeterminationofsebygc-ecd
PubChem CID 2724285
IUPAC Name 3,5-dibromobenzene-1,2-diamine;hydrochloride
SMILES C1=C(C=C(C(=C1Br)N)N)Br.Cl

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 238°C
Color Beige
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula Br2C6H2(NH2)2·HCl
Quantity 500 mg
Beilstein 13, 29
Formula Weight 302.4
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 3, 5-Dibromo-o-phenylenediamine monohydrochloride, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.