missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dibromobenzaldehyde, 98%

Product Code. 11487176
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11487176 5 g 5g
11477176 1 g 1g
11339529 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11487176 Supplier Thermo Scientific Alfa Aesar Supplier No. B25495.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 56990-02-4 | C7H4Br2O | 263.916 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56990-02-4
Molecular Formula C7H4Br2O
Molecular Weight (g/mol) 263.916
MDL Number MFCD00156887
InChI Key ZLDMZIXUGCGKMB-UHFFFAOYSA-N
Synonym 3,5-dibromo-benzaldehyde, benzaldehyde, 3,5-dibromo, pubchem3069, acmc-209lvd, ksc493o2p, 3,5-bis bromanyl benzaldehyde, paragos 530307, 3,5-dibromo-phenyl-methanone
PubChem CID 622077
IUPAC Name 3,5-dibromobenzaldehyde
SMILES C1=C(C=C(C=C1Br)Br)C=O

Specifications

Melting Point 89°C to 93°C
Quantity 5 g
Beilstein 2573432
Sensitivity Air Sensitive
Formula Weight 263.93
Percent Purity 98%
Chemical Name or Material 3,5-Dibromobenzaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.