missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dibromobenzamide, 97%

Product Code. 11471477
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11471477 25 g 25g
11451477 1 g 1g
11461477 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11471477 Supplier Thermo Scientific Alfa Aesar Supplier No. B24946.14
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 175205-85-3 | C7H5Br2NO | 278.931 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 175205-85-3
Molecular Formula C7H5Br2NO
Molecular Weight (g/mol) 278.931
MDL Number MFCD00178771
InChI Key IOFGMNBDIDEBIQ-UHFFFAOYSA-N
Synonym 3,5-dibromo-benzoic acid amide, acmc-20andi, 3,5-dibromo-benzamide, maybridge1_007750, 3,5-bis bromanyl benzamide, 3,5-dibromobenzamide
PubChem CID 2735937
IUPAC Name 3,5-dibromobenzamide
SMILES C1=C(C=C(C=C1Br)Br)C(=O)N

Specifications

Melting Point 180°C to 184°C
Quantity 25 g
Formula Weight 278.93
Percent Purity 97%
Chemical Name or Material 3,5-Dibromobenzamide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.