missing translation for 'onlineSavingsMsg'
Learn More

3,5-dibromobenzamide, Thermo Scientific™

Product Code. 10032504
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10032504 1 g 1g
10452464 10 g 10g
10683445 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10032504 Supplier Thermo Scientific Maybridge Supplier No. SEW04141DA
 Restricted Item
Estimated Shipment: 16-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 175205-85-3
Molecular Formula C7H5Br2NO
Molecular Weight (g/mol) 278.931
InChI Key IOFGMNBDIDEBIQ-UHFFFAOYSA-N
Synonym 3,5-dibromo-benzoic acid amide, acmc-20andi, 3,5-dibromo-benzamide, maybridge1_007750, 3,5-bis bromanyl benzamide, 3,5-dibromobenzamide
PubChem CID 2735937
IUPAC Name 3,5-dibromobenzamide
SMILES C1=C(C=C(C=C1Br)Br)C(=O)N

Specifications

Assay Percent Range 97%
Quantity 1 g
Formula Weight 278.93
Percent Purity 97%
Chemical Name or Material 3, 5-Dibromobenzamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.