missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dichloro-2-hydroxybenzenesulfonic acid sodium salt, 98%

Product Code. 11479602
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11479602 10 g 10g
11489602 50 g 50g
11499602 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11479602 Supplier Thermo Scientific Alfa Aesar Supplier No. A10732.09
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 54970-72-8 | C6H3Cl2NaO4S | 265.036 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54970-72-8
Molecular Formula C6H3Cl2NaO4S
Molecular Weight (g/mol) 265.036
MDL Number MFCD00009798
InChI Key NMWCVZCSJHJYFW-UHFFFAOYSA-M
Synonym sodium 3,5-dichloro-2-hydroxybenzenesulfonate, dhbs, 3,5-dichloro-2-hydroxybenzenesulfonic acid sodium salt, sodium 3,5-dichloro-2-hydroxybenzenesulphonate, dchbs, 3,5-dichloro-2-hydroxybenzenesulfonic acid, sodium salt, 2,4-dichlorophenol-6-sulfonic acid sodium salt, sodium 3,5-chloro-6-hydroxybenzenesulfonate, benzenesulfonic acid, 3,5-dichloro-2-hydroxy-, monosodium salt, 3,5-dichloro-2-hydroxybenzenesulphonic acid sodium salt
PubChem CID 3085077
IUPAC Name sodium;3,5-dichloro-2-hydroxybenzenesulfonate
SMILES C1=C(C=C(C(=C1Cl)O)S(=O)(=O)[O-])Cl.[Na+]

Specifications

Melting Point >300°C
Quantity 10 g
Beilstein 3640643
Formula Weight 265.05
Percent Purity 99%
Chemical Name or Material 3,5-Dichloro-2-hydroxybenzenesulfonic acid sodium salt
compliance-icons
Product Identifier
  • 3,5-Dichloro-2-hydroxybenzenesulphonic acid sodium salt
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P362-Take off contaminated clothing.
Supplemental information
  • MIXTURE LIST-Contain: 3,5-Dichloro-2-hydroxybenzenesulphonic acid sodium salt

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.