missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dichloro-2-hydroxybenzenesulfonyl chloride, 97%

Product Code. 11388635
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11388635 10 g 10g
11398635 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11388635 Supplier Thermo Scientific Alfa Aesar Supplier No. L12147.09
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 23378-88-3 | C6H3Cl3O3S | 261.50 g/mol

3,5-Dichloro-2-hydroxybenzenesulfonyl chloride is used in the synthesis of (1R,2R)-(+)-1,2-(3,3?,5,5?-tetrachloro-2,2?-dihydroxydibenzenesulfonamido)-1,2-diphenylethane1 and (1R,2R)-(+)- 1,2-(3,3?,5,5?-tetrachloro-2,2?-dihydroxydibenzenesulfonamido)cyclohexane. It was used as chromogenic system in one-step kinetic method for the determination of 5?-nucleotidase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Dichloro-2-hydroxybenzenesulfonyl chloride is used in the synthesis of (1R,2R)-(+)-1,2-(3,3′,5,5′-tetrachloro-2,2′-dihydroxydibenzenesulfonamido)-1,2-diphenylethane1 and (1R,2R)-(+)- 1,2-(3,3′,5,5′-tetrachloro-2,2′-dihydroxydibenzenesulfonamido)cyclohexane. It was used as chromogenic system in one-step kinetic method for the determination of 5′-nucleotidase.

Solubility
Hydrolyzes in water.

Notes
Moisture Sensitive. Incompatible with bases, moisture and oxidizing agents. Store under dry inert gas. Protect from humidity and water.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23378-88-3
Molecular Formula C6H3Cl3O3S
Molecular Weight (g/mol) 261.50
MDL Number MFCD00007432
InChI Key KXFQRJNVGBIDHA-UHFFFAOYSA-N
Synonym 3,5-dichloro-2-hydroxybenzenesulphonyl chloride, benzenesulfonyl chloride, 3,5-dichloro-2-hydroxy, 3,5-dichloro-2-hydroxybenzenesulfonylchloride, 3,5-dichloro-2-hydroxybenzene-1-sulfonyl chloride, 3,5-dichloro-2-hydroxy-benzenesulfonyl chloride, acmc-20andd, kxfqrjnvgbidha-uhfffaoysa, 2,4-dichloro-6-chlorosulfonyl phenol, 2,4-dichlorophenol-6-sulfonyl chloride, 3,5-dichloro-2-hydroxyphenyl chlorosulfone
PubChem CID 90078
IUPAC Name 3,5-dichloro-2-hydroxybenzenesulfonyl chloride
SMILES OC1=C(Cl)C=C(Cl)C=C1S(Cl)(=O)=O

Specifications

Melting Point 78°C to 80°C
Quantity 10 g
UN Number UN3261
Beilstein 2940993
Formula Weight 261.51
Percent Purity 97%
Chemical Name or Material 3,5-Dichloro-2-hydroxybenzenesulfonyl chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.