missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dichloro-4-(2-chloro-4-nitrophenoxy)benzene-1-sulfonyl chloride, 97%, Thermo Scientific™

Product Code. 10532452
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10532452 1 g Amber Glass Bottle 1g
10783033 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10532452 Supplier Thermo Scientific Maybridge Supplier No. BTB03983DA
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 175135-06-5
Molecular Formula C12H5Cl4NO5S
Molecular Weight (g/mol) 417.034
MDL Number MFCD00052029
InChI Key YNPAAILIMIGQIH-UHFFFAOYSA-N
Synonym 3,5-dichloro-4-2-chloro-4-nitrophenoxy benzene-1-sulfonyl chloride, 3,5-dichloro-4-2-chloro-4-nitrophenoxy benzenesulfonyl chloride, 4-2-chloro-4-nitrophenoxy-3,5-dichloro-benzenesulfonyl chloride, 4-2-chloro-4-nitrophenoxy-3,5-dichlorobenzenesulfonyl chloride, benzenesulfonylchloride, 3,5-dichloro-4-2-chloro-4-nitrophenoxy
PubChem CID 2774361
IUPAC Name 3,5-dichloro-4-(2-chloro-4-nitrophenoxy)benzenesulfonyl chloride
SMILES C1=CC(=C(C=C1[N+](=O)[O-])Cl)OC2=C(C=C(C=C2Cl)S(=O)(=O)Cl)Cl

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 417.05
Percent Purity 97%
Chemical Name or Material 3, 5-Dichloro-4-(2-chloro-4-nitrophenoxy)benzene-1-sulfonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.