missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dichloro-4-hydroxybenzenesulfonyl chloride, 97%, Thermo Scientific™

Product Code. 11398996
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11398996

Brand: Thermo Scientific Alfa Aesar L20174.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is employed in the convergent syntheses involving hydroxyarenesulfonyl chlorides.

Solubility
Hydrolyzes in water.

Notes
Moisture sensitive. Store away from water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13432-81-0
Molecular Formula C6H3Cl3O3S
Molecular Weight (g/mol) 261.50
MDL Number MFCD00171551
InChI Key AELFLPJWVRGXKU-UHFFFAOYSA-N
PubChem CID 2735994
SMILES OC1=C(Cl)C=C(C=C1Cl)S(Cl)(=O)=O

Specifications

Melting Point 118°C to 119°C
Quantity 1 g
UN Number UN3261
Beilstein 2806977
Sensitivity Moisture sensitive
Synonym 3,5-dichloro-4-hydroxybenzene-1-sulfonyl chloride, 3,5-dichloro-4-hydroxybenzenesulphonyl chloride, 3,5-dichloro-4-hydroxy benzene sulphonyl chloride, 3,5-dichloro-4-hydroxy-benzenesulfonyl chloride, acmc-1c20j, 3,5-dichloro-4-hydroxyphenyl chlorosulfone, 3,5-dichloro-4-hydroxybenzenesulphonylchloride, benzenesulfonylchloride,3,5-dichloro-4-hydroxy, benzenesulfonyl chloride, 3,5-dichloro-4-hydroxy, benzenesulfonylchloride, 3,5-dichloro-4-hydroxy
Solubility Information Hydrolyzes in water.
IUPAC Name 3,5-dichloro-4-hydroxybenzene-1-sulfonyl chloride
Formula Weight 261.51
Percent Purity 97%
Chemical Name or Material 3,5-Dichloro-4-hydroxybenzenesulfonyl chloride
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.