missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dichlorobenzenethiol, 97%

Product Code. 11975751 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11975751 5 g Glass bottle 5g
11965751 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11975751 Supplier Thermo Scientific Acros Supplier No. 435680050
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 17231-94-6 | C6H4Cl2S | 179.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17231-94-6
Molecular Formula C6H4Cl2S
Molecular Weight (g/mol) 179.07
InChI Key WRXIPCQPHZMXOO-UHFFFAOYSA-N
Synonym 3,5-dichlorothiophenol, 3,5-dichloro thiophenol, 3,5-dichlorobenzenthiol, 3,5-dichlorobenzene-1-thiol, pubchem6804, 3,5-dichloro-thiophenol, 3,5-dichloro-benzenethiol, acmc-209e4e, 3,5-dichlorophenyl mercaptan, benzenethiol, 3,5-dichloro
PubChem CID 2736096
IUPAC Name 3,5-dichlorobenzenethiol
SMILES C1=C(C=C(C=C1Cl)Cl)S

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 62°C to 65°C
Color White to Yellow
Boiling Point 270°C to 272°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula Cl2C6H3SH
Quantity 5 g
Solubility Information Solubility in water: insoluble
Formula Weight 179.07
Percent Purity 97%
Physical Form Crystalline Powder or Solid
Chemical Name or Material 3, 5-Dichlorobenzenethiol
Show More Show Less
compliance-icons
Identifiant produit
  • 3,5-Dichlorobenzenethiol
Mention d'avertissement
  • Warning
Catégorie de danger
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Mention de danger
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Conseils de prudence
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.