missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dichlorobenzyl alcohol, 98%

Product Code. 11465633
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11465633 25 g 25g
11455633 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11465633 Supplier Thermo Scientific Alfa Aesar Supplier No. A13527.14
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 60211-57-6 | C7H6Cl2O | 177.024 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60211-57-6
Molecular Formula C7H6Cl2O
Molecular Weight (g/mol) 177.024
MDL Number MFCD00004634
InChI Key VSNNLLQKDRCKCB-UHFFFAOYSA-N
PubChem CID 43236
IUPAC Name (3,5-dichlorophenyl)methanol
SMILES C1=C(C=C(C=C1Cl)Cl)CO

Specifications

Melting Point 76°C to 80°C
Quantity 25 g
Beilstein 1936389
Synonym 3,5-dichlorobenzyl alcohol, 3,5-dichlorophenyl methanol, benzenemethanol, 3,5-dichloro, 3,5-dichlorobenzylalcohol, 3,5-dichlorobenzylic alcohol, 3,5-dichlorobenzenemethanol, 3,5-dicholorbenzylalcohol, 3,5-dichlorophenyl methan-1-ol, 3, 5-dichlorobenzyl alcohol, pubchem4908
Formula Weight 177.03
Percent Purity 98%
Chemical Name or Material 3,5-Dichlorobenzyl alcohol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.