missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dichlorophenol, 98%

Product Code. 11309529
Change view
Click to view available options
Quantity:
5 g
25 g
50 g
Unit Size:
25g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11309529 5 g 5g
11319529 25 g 25g
11488226 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11309529 Supplier Thermo Scientific Alfa Aesar Supplier No. A15328.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 591-35-5 | C6H4Cl2O | 162.997 g/mol

3,5-Dichlorophenol is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Dichlorophenol is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water. Very soluble in ethanol, ethyl ether and petroleum ether.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 591-35-5
Molecular Formula C6H4Cl2O
Molecular Weight (g/mol) 162.997
MDL Number MFCD00002259
InChI Key VPOMSPZBQMDLTM-UHFFFAOYSA-N
Synonym phenol, 3,5-dichloro, 3,5-dichloro-phenol, unii-fg32l88ko9, 3,5 dichlorophenol, ccris 5905, 3,5-dichloro phenol, pubchem3699, acmc-209mar, dsstox_cid_5006, dsstox_rid_77624
PubChem CID 11571
IUPAC Name 3,5-dichlorophenol
SMILES C1=C(C=C(C=C1Cl)Cl)O

Specifications

Melting Point 63°C to 68°C
Boiling Point 233°C
Odor Phenol-like
Quantity 5 g
UN Number UN2020
Beilstein 1906458
Solubility Information Slightly soluble in water. Very soluble in ethanol,ethyl ether and petroleum ether.
Formula Weight 163
Percent Purity 98%
Chemical Name or Material 3,5-Dichlorophenol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.