missing translation for 'onlineSavingsMsg'
Learn More

(3,5-dichlorophenyl)methanol, Thermo Scientific™

Product Code. 10576714
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10576714 50 g 50g
10109702 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10576714 Supplier Thermo Scientific Maybridge Supplier No. SPB00433EE
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 60211-57-6
Molecular Formula C7H6Cl2O
Molecular Weight (g/mol) 177.024
InChI Key VSNNLLQKDRCKCB-UHFFFAOYSA-N
Synonym 3,5-dichlorobenzyl alcohol, 3,5-dichlorophenyl methanol, benzenemethanol, 3,5-dichloro, 3,5-dichlorobenzylalcohol, 3,5-dichlorobenzylic alcohol, 3,5-dichlorobenzenemethanol, 3,5-dicholorbenzylalcohol, 3,5-dichlorophenyl methan-1-ol, 3, 5-dichlorobenzyl alcohol, pubchem4908
PubChem CID 43236
IUPAC Name (3,5-dichlorophenyl)methanol
SMILES C1=C(C=C(C=C1Cl)Cl)CO

Specifications

Assay Percent Range 97+%
Packaging Amber Glass Bottle
Linear Formula Cl2C6H3CH2OH
Quantity 50 g
Formula Weight 177.03
Percent Purity 97%
Chemical Name or Material (3, 5-Dichlorophenyl)methanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.