missing translation for 'onlineSavingsMsg'
Learn More

3,5-Difluoro-4-methoxyaniline, 97%

Product Code. 11346869
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11346869 1 g 1g
11356869 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11346869 Supplier Thermo Scientific Alfa Aesar Supplier No. H26778.03
 Restricted Item
Estimated Shipment: 22-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 363-47-3 | C7H7F2NO | 159.14 g/mol

3,5-Difluoro-4-methoxyaniline is used in conjunction with thermal energy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Difluoro-4-methoxyaniline is used in conjunction with thermal energy.

Solubility
Insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 363-47-3
Molecular Formula C7H7F2NO
Molecular Weight (g/mol) 159.14
MDL Number MFCD04115910
InChI Key POVSDXPEJZMSEJ-UHFFFAOYSA-N
Synonym 4-amino-2,6-fluoroanisole, benzenamine, 3,5-difluoro-4-methoxy-9ci, 3,5-difluoro-4-methoxy-aniline, benzenamine,3,5-difluoro-4-methoxy, pubchem10104, 3,5-difluoro-4-methxoyaniline, 3,5-difluoro-4-methoxybenzenamine, 3,5-difluoro-4-methoxyphenylamine, 3,5-difluoro-4-methoxybenzeneamine, 3,5-difluoro-4-methoxy-benzenamine
PubChem CID 2783137
IUPAC Name 3,5-difluoro-4-methoxyaniline
SMILES COC1=C(F)C=C(N)C=C1F

Specifications

Melting Point 77°C to 80°C
Odor Characteristic
Quantity 1 g
UN Number UN2811
Solubility Information Insoluble in water.
Formula Weight 159.14
Percent Purity 97%
Chemical Name or Material 3,5-Difluoro-4-methoxyaniline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.