missing translation for 'onlineSavingsMsg'
Learn More

3,5-Difluoro-4-(trifluoromethyl)bromobenzene, Thermo Scientific™

Product Code. 10179649
Change view
Click to view available options
Quantity:
5 g
Packaging:
Amber Glass Bottle
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10179649 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10179649 Supplier Thermo Scientific Maybridge Supplier No. MAY00121DE
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
This item is not returnable. View return policy

Specifications

CAS 156243-64-0
Packaging Amber Glass Bottle
Molecular Formula C7H2BrF5
Quantity 5 g
Synonym 3,5-difluoro-4-trifluoromethyl bromobenzene, 5-bromo-1,3-difluoro-2-trifluoromethyl benzene, 4-bromo-2,6-difluorobenzotrifluoride, benzene, 5-bromo-1,3-difluoro-2-trifluoromethyl, acmc-1cioh, intermediates-zcf02186, 2,6-difluoro-4-bromobenzotrifluoride, 1-bromo-3,5-difluoro-4-trifluoromethylbenzene, 3,5-difluoro-4-trifluoromethylbromobenzene, benzene,5-bromo-1,3-difluoro-2-trifluoromethyl
InChI Key QPJKIRNIIXIPIE-UHFFFAOYSA-N
SMILES C1=C(C=C(C(=C1F)C(F)(F)F)F)Br
IUPAC Name 5-bromo-1,3-difluoro-2-(trifluoromethyl)benzene
Molecular Weight (g/mol) 260.989
PubChem CID 23450919
Formula Weight 260.99
Percent Purity 97%
Chemical Name or Material 3, 5-difluoro-4-(trifluoromethyl)bromobenzene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.