missing translation for 'onlineSavingsMsg'
Learn More

3,5-Difluoroaniline, 98%

Product Code. 10607831 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
1g
50g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 10607831

Brand: Thermo Scientific Acros 311420010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

CAS: 372-39-4 | C6H5F2N | 129.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 372-39-4
Molecular Formula C6H5F2N
Molecular Weight (g/mol) 129.11
MDL Number MFCD00007763
InChI Key KQOIBXZRCYFZSO-UHFFFAOYSA-N
Synonym benzenamine, 3,5-difluoro, 3,5-difluorobenzenamine, 3,5-difluorophenylamine, 5an, 3,5-difluoranilin, 3,5-difluoroanilin, 3,5 difluoroaniline, 3.5-difluoroaniline, pubchem2296, 3, 5-difluoroaniline
PubChem CID 96595
IUPAC Name 3,5-difluoroaniline
SMILES C1=C(C=C(C=C1F)F)N

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 37°C to 41°C
Color White to Yellow
Density 75g/mL
Boiling Point 82°C (20.0 mmHg)
Flash Point 75°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula F2C6H3NH2
Quantity 1 g
Formula Weight 129.11
Percent Purity 98%
Physical Form Low Melting Solid
Chemical Name or Material 3, 5-Difluoroaniline
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.