missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3,5-Difluorobenzeneboronic acid neopentyl glycol ester, 98+%

Product Code. 11304016
Click to view available options
Quantity:
5 g
Unit Size:
5g
This item is not returnable. View return policy

Product Code. 11304016

Brand: Alfa Aesar™ L17266.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 216393-57-6
Molecular Formula C11H13BF2O2
Molecular Weight (g/mol) 226.03
MDL Number MFCD01318139
InChI Key OLHRAEPCGZMMTI-UHFFFAOYSA-N
Synonym 2-3,5-difluorophenyl-5,5-dimethyl-1,3,2-dioxaborinane, 3,5-difluorobenzeneboronic acid neopentyl glycol ester, 3,5-difluorophenylboronic acid neopentyl glycol ester, 3,5-difluorobenzeneboronic acid neopentyl glycol cyclic ester, 2-3,5-difluorophenyl-5,5-dimethyl-1,3,2-dioxaborane, 2-3,5-difluorophenyl-5,5-dimethyl-1,3,2-dioxaborine, 1,3,2-dioxaborinane, 2-3,5-difluorophenyl-5,5-dimethyl, 3,5-difluorobenzenebornoic acid neopentyl glycol cyclic ester, 2-3,5-difluorophenyl-5,5-dimethyldihydro-4h-1,3,2-dioxaborin
PubChem CID 2774056
IUPAC Name 2-(3,5-difluorophenyl)-5,5-dimethyl-1,3,2-dioxaborinane
SMILES B1(OCC(CO1)(C)C)C2=CC(=CC(=C2)F)F

Specifications

Melting Point 43°C to 45°C
Quantity 5 g
Formula Weight 226.03
Percent Purity ≥98%
Chemical Name or Material 3,5-Difluorobenzeneboronic acid neopentyl glycol ester
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.