missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,5-Difluorophenol, 98+%
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 51°C to 55°C |
| Boiling Point | 65°C to 68°C (1 mmHg) |
| Flash Point | 70°C (158°F) |
| Odor | Phenol-like |
| Quantity | 5 g |
| UN Number | UN3261 |
| Beilstein | 2078616 |
| Synonym | phenol, 3,5-difluoro, 3,5-difluoro phenol, phenol,3,5-difluoro, 3,5difluorophenol, 3,5-difluorphenol, 3.5-difluorophenol, 3,5-difluoro-phenol, phenol derivative, 6, pubchem1496, acmc-209gvc |
| Formula Weight | 130.09 |
| Percent Purity | ≥98% |
| Show More |
Safety and Handling
Product Identifier
- 3,5-Difluorophenol
- Danger
- Flammable solid Category 2
- Skin corrosion/irritation Category 1 B
- H228-Flammable solid.
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H314-Causes severe skin burns and eye damage.
- H332-Harmful if inhaled.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P233-Keep container tightly closed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contains : 3,5-Difluorophenol
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction