missing translation for 'onlineSavingsMsg'
Learn More

3,5-Difluorophenol, 98+%

Product Code. 11462178
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11462178 25 g 25g
11442178 1 g 1g
11452178 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11462178 Supplier Thermo Scientific Alfa Aesar Supplier No. A15177.14
 Restricted Item
Estimated Shipment: 23-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 2713-34-0 | C6H4F2O | 130.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2713-34-0
Molecular Formula C6H4F2O
Molecular Weight (g/mol) 130.09
MDL Number MFCD00002255
InChI Key HJSSBIMVTMYKPD-UHFFFAOYSA-N
PubChem CID 75928
IUPAC Name 3,5-difluorophenol
SMILES OC1=CC(F)=CC(F)=C1

Specifications

Melting Point 51°C to 55°C
Boiling Point 65°C to 68°C (1 mmHg)
Flash Point 70°C (158°F)
Odor Phenol-like
Quantity 25 g
UN Number UN3261
Beilstein 2078616
Synonym phenol, 3,5-difluoro, 3,5-difluoro phenol, phenol,3,5-difluoro, 3,5difluorophenol, 3,5-difluorphenol, 3.5-difluorophenol, 3,5-difluoro-phenol, phenol derivative, 6, pubchem1496, acmc-209gvc
Formula Weight 130.09
Percent Purity ≥98%
Chemical Name or Material 3,5-Difluorophenol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.