missing translation for 'onlineSavingsMsg'
Learn More

3,5-Difluorophenylacetic acid, 99%, ACROS Organics™

Product Code. 10297560 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
1g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10297560 1g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10297560 Supplier Thermo Scientific Acros Supplier No. 307790010
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 105184-38-1
CAS Min % 98.5
CAS Max % 100.0
Melting Point 66°C to 69°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Molecular Formula C8H6F2O2
Linear Formula F2C6H3CH2CO2H
MDL Number MFCD00010316
Quantity 1 g
Synonym 3,5-difluorophenylacetic acid, 2-3,5-difluorophenyl acetic acid, 3,5-difluorophenylaceticacid, 3,5-difluorobenzeneacetic acid, 3,5-difluorophenyl acetic acid, 3,5-difluoro-phenyl-acetic acid, benzeneacetic acid, 3,5-difluoro, 3,5-diifluorophenylacetic acid, 3,5-difluorophenylac
InChI Key IGGNSAVLXJKCNH-UHFFFAOYSA-N
SMILES C1=C(C=C(C=C1F)F)CC(=O)O
IUPAC Name 2-(3,5-difluorophenyl)acetic acid
Molecular Weight (g/mol) 172.131
PubChem CID 145424
Formula Weight 172.13
Percent Purity ≥98.5%
Physical Form Crystalline Powder
Chemical Name or Material 3, 5-Difluorophenylacetic acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.