missing translation for 'onlineSavingsMsg'
Learn More

3,5-Difluorophenylhydrazine hydrochloride, 97%

Product Code. 11386589
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11386589 5 g 5g
11376589 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11386589 Supplier Thermo Scientific Alfa Aesar Supplier No. H26189.06
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 502496-27-7 | C6H7ClF2N2 | 180.583 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 502496-27-7
Molecular Formula C6H7ClF2N2
Molecular Weight (g/mol) 180.583
MDL Number MFCD03094171
InChI Key CKCNKYAHVKNKHQ-UHFFFAOYSA-N
Synonym 3,5-difluorophenylhydrazine hydrochloride, 3,5-difluorophenyl hydrazine hydrochloride, 3,5-difluorophenylhydrazine hcl, 3,5-difluorophenylhydrazinehydrochloride, 1-3,5-difluorophenyl hydrazine hydrochloride, 3,5-difluoro phenylhydrazine, hydrazine, 3,5-difluorophenyl-, monohydrochloride, pubchem1085, pubchem20776, c6h6f2n2.hcl
PubChem CID 16217172
IUPAC Name (3,5-difluorophenyl)hydrazine;hydrochloride
SMILES C1=C(C=C(C=C1F)F)NN.Cl

Specifications

Melting Point 264°C to 266°C
Quantity 5 g
Formula Weight 180.59
Percent Purity 97%
Chemical Name or Material 3,5-Difluorophenylhydrazine hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.