missing translation for 'onlineSavingsMsg'
Learn More

3,5-Difluorophenylmagnesium bromide, 0.50M in 2-MeTHF

Product Code. 15461406
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15461406 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15461406 Supplier Thermo Scientific Alfa Aesar Supplier No. H54622.AE
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 62351-47-7 | C6H3BrF2Mg | 217.30 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 62351-47-7
Molecular Formula C6H3BrF2Mg
MDL Number MFCD00672005
Quantity 100 mL
UN Number UN2924
Sensitivity Air and moisture sensitive; May form a precipitate
Synonym 3,5-difluorophenylmagnesium bromide, bromo 3,5-difluorophenyl magnesium, 3,5-difluorophenylmagnesiumbromide, curgxpmircwedb-uhfffaoysa-m, 3,5-difluorophenyl magnesium bromide, 3,5-difluorophenylmagnesium bromide, 0.50m in 2-methf, 3,5-difluorophenylmagnesium bromide 0.5 m in tetrahydrofuran, 3,5-difluorophenylmagnesium bromide solution, 0.5 m in thf, grignard reagent
InChI Key CURGXPMIRCWEDB-UHFFFAOYSA-M
SMILES FC1=CC([Mg]Br)=CC(F)=C1
IUPAC Name magnesium;1,3-difluorobenzene-5-ide;bromide
Molecular Weight (g/mol) 217.30
PubChem CID 2778376
Formula Weight 217.29
Concentration or Composition (by Analyte or Components) 0.50M in 2-MeTHF
Chemical Name or Material 3,5-Difluorophenylmagnesium bromide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.