missing translation for 'onlineSavingsMsg'
Learn More

3',5-Dihydroxy-4',6,7-trimethoxyflavone, 97%

Product Code. 11393546
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11393546 25 mg 25mg
11303556 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11393546 Supplier Thermo Scientific Alfa Aesar Supplier No. L14495.MB
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 855-96-9 | C18H16O7 | 344.319 g/mol

3',5-Dihydroxy-4',6,7-trimethoxyflavone is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3′,5-Dihydroxy-4′,6,7-trimethoxyflavone is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

Solubility
Insoluble in water. Soluble in chloroform and methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 855-96-9
Molecular Formula C18H16O7
Molecular Weight (g/mol) 344.319
MDL Number MFCD00016929
InChI Key KLAOKWJLUQKWIF-UHFFFAOYSA-N
Synonym eupatorin, 3',5-dihydroxy-4',6,7-trimethoxyflavone, eupatorine, 5-hydroxy-2-3-hydroxy-4-methoxyphenyl-6,7-dimethoxy-4h-chromen-4-one, unii-3j474av6my, 5-hydroxy-2-3-hydroxy-4-methoxyphenyl-6,7-dimethoxy-4h-1-benzopyran-4-one, 6-methoxyluteolin 4',7-dimethyl ether, 5-hydroxy-2-3-hydroxy-4-methoxyphenyl-6,7-dimethoxychromen-4-one, kinome_3214, spectrum_001037
PubChem CID 97214
IUPAC Name 5-hydroxy-2-(3-hydroxy-4-methoxyphenyl)-6,7-dimethoxychromen-4-one
SMILES COC1=C(C=C(C=C1)C2=CC(=O)C3=C(C(=C(C=C3O2)OC)OC)O)O

Specifications

Melting Point 190°C to 192°C
Quantity 25 mg
Beilstein 1355109
Merck Index 14,3901
Solubility Information Insoluble in water. Soluble in chloroform and methanol.
Formula Weight 344.32
Percent Purity 97%
Chemical Name or Material 3',5-Dihydroxy-4',6,7-trimethoxyflavone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.