missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dihydroxy-4-methylbenzoic acid, 97%

Product Code. 11325259
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11325259 25 g 25g
11305259 1 g 1g
11315259 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11325259 Supplier Thermo Scientific Alfa Aesar Supplier No. A19196.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 28026-96-2 | C8H7O4 | 167.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 28026-96-2
Molecular Formula C8H7O4
Molecular Weight (g/mol) 167.14
MDL Number MFCD00002514
InChI Key KMRRXSZDSGYLCD-UHFFFAOYSA-M
PubChem CID 2748045
IUPAC Name 3,5-dihydroxy-4-methylbenzoic acid
SMILES CC1=C(O)C=C(C=C1O)C([O-])=O

Specifications

Melting Point 266°C to 207°C
Quantity 25 g
Synonym 3,5-dihydroxy-p-toluic acid, benzoic acid, 3,5-dihydroxy-4-methyl, 4-methyl-3,5-dihydroxy benzoic acid, 3,5-dihydroxy-4-methyl-benzoic acid, pubchem3868, acmc-20a50j, paragos 440034, 3,5-dihydroxy4-methylbenzoic acid, 3,5-dihydroxy-4-methyl benzoic acid, benzoic acid,3,5-dihydroxy-4-methyl
Formula Weight 168.15
Percent Purity 97%
Chemical Name or Material 3,5-Dihydroxy-4-methylbenzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.