missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dihydroxybenzoic acid, 97%

Product Code. 10418260 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Unit Size:
100g
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10418260 500 g 500g
10122860 100 g 100g
10637763 2.5 kg 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10418260 Supplier Thermo Scientific Acros Supplier No. 114905000
 Restricted Item
Estimated Shipment: 22-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 99-10-5 | C7H6O4 | 154.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99-10-5
Molecular Formula C7H6O4
Molecular Weight (g/mol) 154.12
MDL Number MFCD00002512
InChI Key UYEMGAFJOZZIFP-UHFFFAOYSA-N
Synonym alpha-resorcylic acid, 5-carboxyresorcinol, benzoic acid, 3,5-dihydroxy, unii-2wc5lmo6l1, 3,5-dihydroxy benzoic acid, 3,5-dihydroxy-benzoic acid, 3,5-dhba, 3,5-dihydroxybenzoicacid, a-resorcylic acid, .alpha.-resorcylic acid
PubChem CID 7424
ChEBI CHEBI:39912
IUPAC Name 3,5-dihydroxybenzoic acid
SMILES OC(=O)C1=CC(O)=CC(O)=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 235.0°C to 240.0°C
Color White to Beige
Flash Point >200°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Plastic bottle
Linear Formula (HO)2C6H3CO2H
Quantity 500 g
Beilstein 10,404
Solubility Information Solubility in water: 84g/L (20°C). Other solubilities: soluble in ethanol and ether
Formula Weight 154.12
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 3,5-Dihydroxybenzoic acid, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 3,5-Dihydroxybenzoic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.