missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dihydroxytoluene, 99%

Product Code. 11389415
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11389415 25 g 25g
11379415 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11389415 Supplier Thermo Scientific Alfa Aesar Supplier No. L18567.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 504-15-4 | C7H8O2 | 124.14 g/mol

Used in production of orcein

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 504-15-4
Molecular Formula C7H8O2
Molecular Weight (g/mol) 124.14
MDL Number MFCD00002291
InChI Key OIPPWFOQEKKFEE-UHFFFAOYSA-N
Synonym orcinol, 3,5-dihydroxytoluene, 5-methylresorcinol, 1,3-dihydroxy-5-methylbenzene, orcin, 5-methyl-1,3-benzenediol, 5-methylresorcin, 3-hydroxy-5-methylphenol, 3,5-toluenediol, 1,3-benzenediol, 5-methyl
PubChem CID 10436
ChEBI CHEBI:16536
IUPAC Name 5-methylbenzene-1,3-diol
SMILES CC1=CC(O)=CC(O)=C1

Specifications

Melting Point 106°C to 108°C
Boiling Point 287°C to 290°C
Quantity 25 g
Beilstein 1071903
Sensitivity Air Sensitive
Merck Index 14,6864
Formula Weight 124.14
Percent Purity 99%
Chemical Name or Material 3,5-Dihydroxytoluene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.