missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethoxy-4-methylbenzoic acid, 97%

Product Code. 11412547
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11412547 5 g 5g
11422547 25 g 25g
11432547 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11412547 Supplier Thermo Scientific Alfa Aesar Supplier No. A14801.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 61040-81-1 | C10H12O4 | 196.202 g/mol

3,5-Dimethoxy-4-methylbenzoic acid undergoes reduction in the presence of sodium in ethanol to prepare 1,4-dihydro-3,5-dimethoxy- kmethylbenzoic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Dimethoxy-4-methylbenzoic acid undergoes reduction in the presence of sodium in ethanol to prepare 1,4-dihydro-3,5-dimethoxy- kmethylbenzoic acid.

Solubility
Soluble in liquid ammonia.

Notes
Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 61040-81-1
Molecular Formula C10H12O4
Molecular Weight (g/mol) 196.202
MDL Number MFCD00017506
InChI Key QIBMVRYNEXOCCF-UHFFFAOYSA-N
Synonym 3,5-dimethoxy-4-methyl-benzoic acid, benzoic acid,3,5-dimethoxy-4-methyl, 3,5-dimethoxy-4-methylbenzoicacid, acmc-1b58y, 3,5-dimethoxy4-methylbenzoic acid, 4-methyl-3,5-dimethoxybenzoic acid, 3,5-dimethoxy-p-toluic acid, labotest-bb lt00455447, 3,5-dimethoxy-4-methyl benzoic acid, 3,5-di-methoxy-4-methyl-benzoic acid
PubChem CID 3764486
IUPAC Name 3,5-dimethoxy-4-methylbenzoic acid
SMILES CC1=C(C=C(C=C1OC)C(=O)O)OC

Specifications

Melting Point 210°C to 216°C
Quantity 5 g
Beilstein 2695238
Solubility Information Soluble in liquid ammonia.
Formula Weight 196.2
Percent Purity 97%
Chemical Name or Material 3,5-Dimethoxy-4-methylbenzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.